CAS RN 38861-88-0
CAS RN 38861-88-0
IUPAC Name:
4-(2-methylpropyl)benzoic acid
Synonyms:
4-(2-methylpropyl)benzoic acid
Molecular Weight (g/mol):
178.23
Molecular Formula:
C11H14O2
InChi Key:
VUBBCFWWSKOHTH-UHFFFAOYSA-N
SMILES:
CC(C)CC1=CC=C(C=C1)C(O)=O
1
–
4
of
4
results
1
4-Isobutylbenzoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
4-Isobutylbenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Isobutylbenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
4-Isobutylbenzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.