Learn More
4-Isobutylbenzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Isobutylbenzoic Acid |
| CAS | 38861-88-0 |
| Molecular Formula | C11 H14 O2 |
| Quantity | 5 g |
| Synonym | 4-Isobutylbenzoic acid, Benzoic acid, 4-(2-methylpropyl)-, 4-(2-Methylpropyl)benzoic acid, NSC 168984, p-(2-Methylpropyl)benzoic acid, p-Isobutylbenzoic acid |
| InChI Formula | InChI=1S/C11H14O2/c1-8(2)7-9-3-5-10(6-4-9)11(12)13/h3-6,8H,7H2,1-2H3,(H,12,13) |
| SMILES | CC(C)Cc1ccc(cc1)C(=O)O |
| IUPAC Name | 4-(2-methylpropyl)benzoic acid |
| Molecular Weight (g/mol) | 178.23 |
| Formula Weight | 178.0994 |