accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNSOOClNH

CAS RN 364-98-7

IUPAC Name: 7-chloro-3-methyl-4H-1λ⁶,2,4-benzothiadiazine-1,1-dione
Synonyms: hyperstat proglicem mutabase proglycem hypertonalum dizoxide
Molecular Weight (g/mol): 230.67
Molecular Formula: C8H7ClN2O2S
InChi Key: GDLBFKVLRPITMI-UHFFFAOYSA-N
SMILES: CC1=NS(=O)(=O)C2=CC(Cl)=CC=C2N1
Quantity 
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results