accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3482-86-8

CH3NH2NH2HNCH3NH2NH2HNOSOOO

CAS RN 3482-86-8

IUPAC Name: bis(carbamimidoyl(ethyl)azanium) sulfate
Synonyms: bis(carbamimidoyl(ethyl)azanium) sulfate
Molecular Weight (g/mol): 272.32
Molecular Formula: C6H20N6O4S
InChi Key: UKQVDMIAGTYDFN-UHFFFAOYSA-N
SMILES: [O-]S([O-])(=O)=O.CC[NH2+]C(N)=N.CC[NH2+]C(N)=N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results