Learn More
N-Ethylguanidine sulfate, 98%
Product Code. 11439227
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439227 25 g 25g
11429227 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11439227 Supplier Thermo Scientific Alfa Aesar Supplier No. B21315.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
CAS: 3482-86-8 | [CH3CH2NHC(=NH)NH2]2·H2SO4
1-Ethylguanidine sulfate may be used in chemical synthesis and thermal analysis studies. 1-Ethylguanidine sulfate is used as a reactant in the sunthesis of 3-Ethyl-6-methyl-isocytosines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Ethylguanidine sulfate may be used in chemical synthesis and thermal analysis studies. 1-Ethylguanidine sulfate is used as a reactant in the sunthesis of 3-Ethyl-6-methyl-isocytosines.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents,water, moisture.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3482-86-8
Molecular Formula [CH3CH2NHC(=NH)NH2]2·H2SO4
MDL Number MFCD00013130
Specifications
Melting Point ∼245°C (decomposition)
Odor Odorless
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 272.33
Percent Purity 98%
Chemical Name or Material N-Ethylguanidine sulfate