CAS RN 3464-71-9
CAS RN 3464-71-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1R,3r,5S)-3-Hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] Chloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.