Learn More
(1R,3r,5S)-3-Hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] Chloride, Mikromol™
Product Code. 30461217
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461217 4 x 25 mg 100mg
30050431 100 mg 100mg
30051929 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30461217 Supplier Mikromol Supplier No. MM1266.0300254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1R,3r,5S)-3-Hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] Chloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Azoniaspironortropanol chloride
CAS 3464-71-9
Analyte or Component Names Azoniaspironortropanol chloride
Shipping Condition Room Temperature
Molecular Formula C11 H20 N O . Cl
Certifications/Compliance ISO 17025
Synonym Trospium Chloride Imp. C (EP), Trospium Chloride USP Related Compound C, Trospium Chloride USP RC C, 3-hydroxy-Spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] chloride, 3alpha-Hydroxynortropane-8-spiro-1'-pyrrolidinium chloride, (1alpha,3beta,5alpha)-3-Hydroxy-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium] chloride (1:1), (1R,3r,5S)-3-hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium], Trospium Imp. C, Trospium RC C (USP), Azoniaspironortropanol chloride, Trospium Related Compound C as Chloride, Trospium Chloride Impurity C as Chloride, Trospium Impurity C as Chloride
InChI Formula InChI=1S/C11H20NO.ClH/c13-11-7-9-3-4-10(8-11)12(9)5-1-2-6-12;/h9-11,13H,1-8H2;1H/q+1;/p-1/t9-,10+,11+;
Solution Type Neat
SMILES [Cl-].O[C@H]1C[C@H]2CC[C@@H](C1)[N+]23CCCC3
IUPAC Name (1S,5R)-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-azolidin-1-ium]-3-ol;chloride
Molecular Weight (g/mol) 217.74
Quantity 4 x 25 mg
Formula Weight 217.1233 g/mol