accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OHOSO

CAS RN 33005-95-7

IUPAC Name: 2-(5-benzoylthiophen-2-yl)propanoic acid
Synonyms: tiaprofenic acid nu-tiaprofenic apo-tiaprofenic novo-tiaprofenic tiaprofenic-300 - tab flanid tiaprofenic-200 - tab tiaprofensaeure apo-tiaprofenic tablets florizel brand of tiaprofenic acid
Molecular Weight (g/mol): 260.31
Molecular Formula: C14H12O3S
InChi Key: GUHPRPJDBZHYCJ-UHFFFAOYNA-N
SMILES: CC(C(O)=O)C1=CC=C(S1)C(=O)C1=CC=CC=C1
Certifications/Compliance 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results