Learn More
Tiaprofenic Acid, LoGiCal
Product Code. 30462280
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30462280
Brand: LoGiCal LGCFOR3322.00
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tiaprofenic acid
CAS 33005-95-7
Analyte or Component Names Tiaprofenic acid
Shipping Condition Room Temperature
Molecular Formula C14 H12 O3 S
Certifications/Compliance ISO 9001
Synonym Tiaprofenic acid, (2RS)-2-(5-Benzoylthiophen-2-yl)propanoic acid
InChI Formula InChI=1S/C14H12O3S/c1-9(14(16)17)11-7-8-12(18-11)13(15)10-5-3-2-4-6-10/h2-9H,1H3,(H,16,17)
Solution Type Neat
SMILES CC(C(=O)O)c1ccc(s1)C(=O)c2ccccc2
IUPAC Name 2-(5-benzoylthiophen-2-yl)propanoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 260.31
Quantity 10 mg
Formula Weight 260.0507 g/mol