accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3279-07-0

CH3H3CCH3OHNOO

CAS RN 3279-07-0

IUPAC Name: 4-tert-butyl-2-nitrophenol
Synonyms: 4-(tert-butyl)-2-nitrophenol
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: IHGNADPMUSNTJW-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(O)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results