Learn More
4-tert-Butyl-2-nitrophenol, TRC
Specifications
Specifications
| Chemical Name or Material | 4-tert-Butyl-2-nitrophenol |
| CAS | 3279-07-0 |
| Molecular Formula | C10H13NO3 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C10H13NO3/c1-10(2,3)7-4-5-9(12)8(6-7)11(13)14/h4-6,12H,1-3H3 |
| SMILES | CC(C)(C)c1ccc(O)c(c1)[N+](=O)[O-] |
| IUPAC Name | 4-tert-butyl-2-nitrophenol |
| Molecular Weight (g/mol) | 195.22 |
| Formula Weight | 195.0895 |