CAS RN 32723-67-4
CAS RN 32723-67-4
IUPAC Name:
4-methoxy-3-methylbenzaldehyde
Synonyms:
3-methyl-4-methoxybenzaldehyde
Molecular Weight (g/mol):
150.18
Molecular Formula:
C9H10O2
InChi Key:
MYLBIQHZWFWSMH-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=O)C=C1C
1
–
2
of
2
results
1
4-Methoxy-3-methylbenzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Methoxy-3-methylbenzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.