Learn More
4-Methoxy-3-methylbenzaldehyde, TRC
Product Code. 30471926
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471926 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30471926 Supplier Toronto Research Chemicals Supplier No. TRCM2633505G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Methoxy-3-methylbenzaldehyde
CAS 32723-67-4
Molecular Formula C9H10O2
Quantity 5 g
Synonym (4-Methoxy-3-methylphenyl)formaldehyde, 3-Methyl-4-methoxybenzaldehyde, 3-Methyl-p-anisaldehyde, 4-Methoxy-3-methylbenzaldehyde, 4-Methoxy-3-tolualdehyde, 4-Methoxy-3-methyl-benzaldehyde
InChI Formula InChI=1S/C9H10O2/c1-7-5-8(6-10)3-4-9(7)11-2/h3-6H,1-2H3
SMILES COc1ccc(C=O)cc1C
IUPAC Name 4-methoxy-3-methylbenzaldehyde
Molecular Weight (g/mol) 150.17
Formula Weight 150.0681