Learn More
4-Methoxy-3-methylbenzaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methoxy-3-methylbenzaldehyde |
| CAS | 32723-67-4 |
| Molecular Formula | C9H10O2 |
| Quantity | 1 g |
| Synonym | (4-Methoxy-3-methylphenyl)formaldehyde, 3-Methyl-4-methoxybenzaldehyde, 3-Methyl-p-anisaldehyde, 4-Methoxy-3-methylbenzaldehyde, 4-Methoxy-3-tolualdehyde, 4-Methoxy-3-methyl-benzaldehyde |
| InChI Formula | InChI=1S/C9H10O2/c1-7-5-8(6-10)3-4-9(7)11-2/h3-6H,1-2H3 |
| SMILES | COc1ccc(C=O)cc1C |
| IUPAC Name | 4-methoxy-3-methylbenzaldehyde |
| Molecular Weight (g/mol) | 150.17 |
| Formula Weight | 150.0681 |