CAS RN 327-74-2
CAS RN 327-74-2
IUPAC Name:
4-amino-3-(trifluoromethyl)benzonitrile
Synonyms:
4-amino-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol):
186.14
Molecular Formula:
C8H5F3N2
InChi Key:
MWLZJOBGDXBMBP-UHFFFAOYSA-N
SMILES:
NC1=C(C=C(C=C1)C#N)C(F)(F)F
1
–
4
of
4
results
1
2-Amino-5-cyanobenzotrifluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Amino-3-(trifluoromethyl)benzonitrile, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
2-Amino-5-cyanobenzotrifluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2-Amino-5-cyanobenzotrifluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.