Learn More
2-Amino-5-cyanobenzotrifluoride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Amino-3-(trifluoromethyl)benzonitrile |
| CAS | 327-74-2 |
| Molecular Formula | C8 H5 F3 N2 |
| Quantity | 10 g |
| Synonym | Benzonitrile, 4-amino-3-(trifluoromethyl)-, m-Tolunitrile, 4-amino-α,α,α-trifluoro- (6CI,8CI), 4-Amino-3-(trifluoromethyl)benzonitrile, 2-Amino-5-cyanobenzotrifluoride, 2-Trifluoromethyl-4-cyanoaniline, 3-Trifluoromethyl-4-aminobenzonitrile, 4-Cyano-2-trifluoromethylaniline |
| InChI Formula | InChI=1S/C8H5F3N2/c9-8(10,11)6-3-5(4-12)1-2-7(6)13/h1-3H,13H2 |
| SMILES | Nc1ccc(cc1C(F)(F)F)C#N |
| IUPAC Name | 4-amino-3-(trifluoromethyl)benzonitrile |
| Molecular Weight (g/mol) | 186.1339 |
| Formula Weight | 186.0405 |