CAS RN 326914-09-4
CAS RN 326914-09-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-[(Z)-(2-oxoindolin-3-ylidene)methyl]-1H-pyrrole-3-carboxamide (Desfluorosunitinib), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.