Learn More
N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-[(Z)-(2-oxoindolin-3-ylidene)methyl]-1H-pyrrole-3-carboxamide (Desfluorosunitinib), Mikromol™
Description
N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-[(Z)-(2-oxoindolin-3-ylidene)methyl]-1H-pyrrole-3-carboxamide (Desfluorosunitinib)
Specifications
Specifications
| Chemical Name or Material | Desfluorosunitinib |
| CAS | 326914-09-4 |
| Analyte or Component Names | Desfluorosunitinib |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H28 N4 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-[(Z)-(2-oxoindolin-3-ylidene)methyl]-1H-pyrrole-3-carboxamide, 1H-Pyrrole-3-carboxamide, N-[2-(diethylamino)ethyl]-5-[(Z)-(1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-2,4-dimethyl-, N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-[(Z)-(2-oxoindolin-3-ylidene)methyl]-1H-pyrrole-3-carboxamide, Desfluorosunitinib |
| InChI Formula | InChI=1S/C22H28N4O2/c1-5-26(6-2)12-11-23-22(28)20-14(3)19(24-15(20)4)13-17-16-9-7-8-10-18(16)25-21(17)27/h7-10,13,24H,5-6,11-12H2,1-4H3,(H,23,28)(H,25,27)/b17-13- |
| SMILES | CCN(CC)CCNC(=O)c1c(C)[nH]c(\C=C\2/C(=O)Nc3ccccc23)c1C |
| IUPAC Name | N-[2-(diethylamino)ethyl]-2,4-dimethyl-5-[(Z)-(2-oxoindolin-3-ylidene)methyl]-1H-pyrrole-3-carboxamide |
| Show More |