CAS RN 32624-67-2
CAS RN 32624-67-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1,2,3,4,4a,9,10,10a-Octahydro-1-methyl-7-(1-methylethyl)-phenanthrene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2,3,4,4a,9,10,10a-Octahydro-1-methyl-7-(1-methylethyl)-phenanthrene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,2,3,4,4a,9,10,10a-Octahydro-1-methyl-7-(1-methylethyl)-phenanthrene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.