Learn More
1,2,3,4,4a,9,10,10a-Octahydro-1-methyl-7-(1-methylethyl)-phenanthrene, TRC
Specifications
Specifications
| Chemical Name or Material | 7-Isopropyl-1-methyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene |
| CAS | 32624-67-2 |
| Molecular Formula | C18 H26 |
| Quantity | 2.5 mg |
| Synonym | 1,2,3,4,4a,9,10,10a-Octahydro-7-isopropyl-1-methylphenanthrene, 1,2,3,4,4a,9,10,10a-Octahydro-1-methyl-7-(1-methylethyl)-phenanthrene |
| InChI Formula | InChI=1S/C18H26/c1-12(2)14-7-10-17-15(11-14)8-9-16-13(3)5-4-6-18(16)17/h7,10-13,16,18H,4-6,8-9H2,1-3H3 |
| SMILES | CC(C)c1ccc2C3CCCC(C)C3CCc2c1 |
| IUPAC Name | 7-isopropyl-1-methyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene |
| Molecular Weight (g/mol) | 242.4 |
| Formula Weight | 242.2035 |