accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32598-13-3

ClClClCl

CAS RN 32598-13-3

IUPAC Name: 3,3',4,4'-tetrachloro-1,1'-biphenyl
Synonyms: 3,3',4,4'-tetrachlorobiphenyl
Molecular Weight (g/mol): 291.98
Molecular Formula: C12H6Cl4
InChi Key: UQMGJOKDKOLIDP-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1Cl)C1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Concentration 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results