Learn More
PCB Mixture for HJ 715-2014 / HJ 743-2015 100 μg/mL in Toluene, Dr. Ehrenstorfer
Product Code. 30314400
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314400 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30314400 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000584TO
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 32598-13-3, 37680-73-2, 32598-14-4, 74472-37-0, 31508-00-6, 65510-44-3
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Toluene