CAS RN 3239-45-0
CAS RN 3239-45-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
(S)-(+)-Fenfluramine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Promotions Available
Tocris Bioscience™ Dexfenfluramine hydrochloride
Tocris Bioscience™ Dexfenfluramine hydrochloride
5-HT reuptake inhibitor; also stimulates 5-HT release
3
(S)-(+)-Fenfluramine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(S)-(+)-Fenfluramine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.