Learn More
(S)-(+)-Fenfluramine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-(+)-Fenfluramine Hydrochloride |
| CAS | 3239-45-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12H16F3N • HCl |
| Quantity | 50 mg |
| Synonym | (αS)-N-Ethyl-α-methyl-3-(trifluoromethyl)benzeneethanamine Hydrochloride, (+)-N-Ethyl-α-methyl-m-(trifluoromethyl)phenethylamine Hydrochloride, Adifax, Adipomin, Glypolix, Isomeride, S5614 HCl, (+)-Fenfluramine Hydrochloride, Dexfenfluramine Hydrochloride |
| InChI Formula | InChI=1S/C12H16F3N.ClH/c1-3-16-9(2)7-10-5-4-6-11(8-10)12(13,14)15;/h4-6,8-9,16H,3,7H2,1-2H3;1H/t9-;/m0./s1 |
| SMILES | C[C@H](NCC)CC1=CC=CC(C(F)(F)F)=C1.Cl |
| Molecular Weight (g/mol) | 231.263646 |
| Percent Purity | >95 |