CAS RN 321-55-1
CAS RN 321-55-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Haloxon, MedChemExpress
MedChemExpress Haloxon is an anti-parasitic agent. Haloxon can be used for the research of infections of Parascaris equorum, Oxyuris equi and Strongylus vulgaris. Haloxon also can be used in control of ascarids and hookworms in domesticated animals in combination with Bidimazium.