Learn More
Haloxon, MedChemExpress
Description
Haloxon is an anti-parasitic agent. Haloxon can be used for the research of infections of Parascaris equorum, Oxyuris equi and Strongylus vulgaris. Haloxon also can be used in control of ascarids and hookworms in domesticated animals in combination with Bidimazium.
Specifications
Specifications
| Chemical Name or Material | Haloxon |
| Physical Form | Solid |
| CAS | 321-55-1 |
| Color | Off-White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C14H14Cl3O6P |
| Solubility Information | DMSO : ≥ 33 mg/mL (79.41 mM) |
| SMILES | O=P(OCCCl)(OC1=CC=C(C(O2)=C1)C(C)=C(Cl)C2=O)OCCCl |
| Molecular Weight (g/mol) | 415.59 |
| Show More |