CAS RN 3195-78-6
CAS RN 3195-78-6
IUPAC Name:
N-ethenyl-N-methylacetamide
Synonyms:
acetamide, N-ethenyl-N-methyl-
Molecular Weight (g/mol):
99.13
Molecular Formula:
C5H9NO
InChi Key:
PNLUGRYDUHRLOF-UHFFFAOYSA-N
SMILES:
CN(C=C)C(C)=O
1
–
2
of
2
results
1
N-Methyl-N-vinylacetamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Methyl-N-vinylacetamide, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality