Learn More
N-Methyl-N-vinylacetamide, Dr. Ehrenstorfer
Product Code. 30323800
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323800 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323800 Supplier Dr. Ehrenstorfer Supplier No. DRECA15147700
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Methyl-N-vinylacetamide
CAS 3195-78-6
Analyte or Component Names N-Methyl-N-vinylacetamide
Shipping Condition +20°C
Molecular Formula C5 H9 N O
Certifications/Compliance ISO 17034
Synonym Acetamide, N-methyl-N-vinyl- (6CI,7CI,8CI), N-Ethenyl-N-methylacetamide, N-Vinyl-N-methylacetamide
InChI Formula InChI=1S/C5H9NO/c1-4-6(3)5(2)7/h4H,1H2,2-3H3
SMILES CN(C=C)C(=O)C
IUPAC Name N-ethenyl-N-methylacetamide
Molecular Weight (g/mol) 99.13
Quantity 100 mg
Formula Weight 99.0684 g/mol