accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3147-75-9

H3CCH3CH3CH3H3COHNNN

CAS RN 3147-75-9

IUPAC Name: 2-(2H-1,2,3-benzotriazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol
Synonyms: octrizole cyasorb 5411 octrizolum seesorb 709 奥克三唑 octrizol viosorb 583 cyasorb UV 5411 sumisorb 340 spectra-sorb UV 5411
Molecular Weight (g/mol): 323.44
Molecular Formula: C20H25N3O
InChi Key: IYAZLDLPUNDVAG-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)C1=CC=C(O)C(=C1)N1N=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results