Learn More
2-Benzotriazolyl-4-tert-octylphenol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(2H-Benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol |
| CAS | 3147-75-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20 H25 N3 O |
| Quantity | 100 mg |
| Synonym | 2-(2H-Benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol, 1268A, 2-(2-Hydroxy-5-t-octylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-5-t-octylphenyl)benzotriazole, 2-(2-Hydroxy-5-tert-octylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole, 2-(2H-Benzotriazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol, 2-(2H-Benzotriazol-2-yl)-4-tert-octylphenol, 2-(2H-Benzotriazole-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol, 2-(2'-Hydroxy-5'-tert-octylphenyl)benzotriazole, 2-(2'-Hydroxy-5'-tert-octylphenyl)benztriazole, 2-(5-t-Octyl-2-hydroxyphenyl)benzotriazole, 2-(5'-tert-Octyl-2'-hydroxyphenyl)benzotriazole, 2-Benzotriazolyl-4-tert-octylphenol, 2-[2'-Hydroxy-5'-(1,1,3,3-tetramethylbutyl)phenyl]benzotriazole, ADK Stab LA 29, BLS 5411, Biosoap 583, Chisorb 5411, Cyasorb 5411, Cyasorb UV 5411, Eversorb 72, Fisorb 329, JF 83, Kemisorb 79, LA 29, Octrizole, Seesorb 709, Seesorb 709G, Songsorb 3290, Spectra-Sorb UV 5411, Sumisorb 340, Tinuvin 329, UV 329, UV 5411, UVA 235, Uvinul 3029, Viosorb 583 |
| InChI Formula | InChI=1S/C20H25N3O/c1-19(2,3)13-20(4,5)14-10-11-18(24)17(12-14)23-21-15-8-6-7-9-16(15)22-23/h6-12,24H,13H2,1-5H3 |
| SMILES | CC(C)(C)CC(C)(C)c1ccc(O)c(c1)n2nc3ccccc3n2 |
| IUPAC Name | 2-(benzotriazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol |
| Molecular Weight (g/mol) | 323.43 |
| Show More |