CAS RN 29700-34-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-[15N]Phenylalanine, TRC
CAS: 29700-34-3 Molecular Formula: C9H1115NO2 Molecular Weight (g/mol): 166.19 Synonym: L-Phenylalanine-15N IUPAC Name: (2S)-2-(^{15}N)azanyl-3-phenylpropanoic acid SMILES: [15NH2][C@@H](Cc1ccccc1)C(=O)O