Learn More
L-[15N]Phenylalanine, TRC
Product Code. 17969164
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17969164 10 mg 10mg
17927444 50 mg 50mg
17938974 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17969164 Supplier Toronto Research Chemicals Supplier No. TRCP33552310MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29700-34-3
Molecular Formula C9H1115NO2
Molecular Weight (g/mol) 166.19
Synonym L-Phenylalanine-15N
IUPAC Name (2S)-2-(^{15}N)azanyl-3-phenylpropanoic acid
SMILES [15NH2][C@@H](Cc1ccccc1)C(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1/i10+1
Formula Weight 166.076
Chemical Name or Material L-[15N]Phenylalanine