accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 294885-63-5

H3CNCH3SOONHOH3C

CAS RN 294885-63-5

IUPAC Name: N-[4-(dipropylsulfamoyl)phenyl]acetamide
Synonyms: N-[4-(dipropylsulfamoyl)phenyl]acetamide
Molecular Weight (g/mol): 298.4
Molecular Formula: C14H22N2O3S
InChi Key: RSKJZTBKLIORJO-UHFFFAOYSA-N
SMILES: CCCN(CCC)S(=O)(=O)C1=CC=C(NC(C)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results