CAS RN 28197-66-2
CAS RN 28197-66-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Acebutolol EP Impurity A, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3’-Acetyl-4’-(2,3-epoxypropoxy)butyranilide, TRC
CAS: 28197-66-2 Molecular Formula: C15 H19 N O4 Molecular Weight (g/mol): 277.32 Synonym: N-[3-Acetyl-4-[(2RS)-oxiran-2-ylmethoxy]phenyl]butanamide,Acebutolol Hydrochloride Imp. A (EP) IUPAC Name: N-[3-acetyl-4-(oxiran-2-ylmethoxy)phenyl]butanamide SMILES: CCCC(=O)Nc1ccc(OCC2CO2)c(c1)C(=O)C