CAS RN 28059-64-5
CAS RN 28059-64-5
IUPAC Name:
2-benzylaniline
Synonyms:
2-benzylaniline
Molecular Weight (g/mol):
183.25
Molecular Formula:
C13H13N
InChi Key:
DWOBGCPUQNFAFB-UHFFFAOYSA-N
SMILES:
NC1=CC=CC=C1CC1=CC=CC=C1
1
–
3
of
3
results
1
2-Benzylaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Benzylaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Benzylaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.