Learn More
2-Benzylaniline, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Benzylaniline |
| CAS | 28059-64-5 |
| Molecular Formula | C13H13N |
| Quantity | 1 g |
| Synonym | 2-(Phenylmethyl)benzenamine, (2-Aminophenyl)phenylmethane, 2-Benzylbenzenamine, o-Aminodiphenylmethane, o-Benzylaniline |
| InChI Formula | InChI=1S/C13H13N/c14-13-9-5-4-8-12(13)10-11-6-2-1-3-7-11/h1-9H,10,14H2 |
| SMILES | Nc1ccccc1Cc2ccccc2 |
| IUPAC Name | 2-benzylaniline |
| Molecular Weight (g/mol) | 183.25 |
| Formula Weight | 183.1048 |