accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 27918-14-5

H2NHNO

CAS RN 27918-14-5

IUPAC Name: 2-amino-9,10-dihydroacridin-9-one
Synonyms: AMAC
Molecular Weight (g/mol): 210.24
Molecular Formula: C13H10N2O
InChi Key: PIGCSKVALLVWKU-UHFFFAOYSA-N
SMILES: NC1=CC=C2NC3=CC=CC=C3C(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results