Learn More
MedChemExpress 2-Aminoacridone
Specifications
Specifications
| Content And Storage | -20°C, protect from light ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Product Type | Dye Reagents |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 2.94 mg/mL (13.98 mM; ultrasonic and warming and heat to 60°C) |
| Molecular Weight (g/mol) | 210.24 g/mol |
| CAS | 27918-14-5 |