CAS RN 27652-89-7
CAS RN 27652-89-7
IUPAC Name:
(4-chlorophenyl)(pyridin-2-yl)methanol
Synonyms:
(4-chlorophenyl)(pyridin-2-yl)methanol
Molecular Weight (g/mol):
219.67
Molecular Formula:
C12H10ClNO
InChi Key:
ZFUPOFQRQNJDNS-UHFFFAOYNA-N
SMILES:
OC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1
1
–
2
of
2
results
2
4-Chlorophenyl-2-pyridinylmethanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.