Learn More
4-Chlorophenyl-2-pyridinylmethanol, TRC
Specifications
Specifications
| CAS | 27652-89-7 |
| Molecular Formula | C12 H10 Cl N O |
| Quantity | 1 g |
| Synonym | α-(4-Chlorophenyl)-2-pyridinemethanol, 2-Pyridinemethanol, α-(p-chlorophenyl)- (7CI,8CI), α-(4-Chlorophenyl)-2-pyridinemethanol, (p-Chlorophenyl)(2-pyridinyl)methanol, NSC 31264, NSC 47970 |
| InChI Formula | InChI=1S/C12H10ClNO/c13-10-6-4-9(5-7-10)12(15)11-3-1-2-8-14-11/h1-8,12,15H |
| SMILES | OC(c1ccc(Cl)cc1)c2ccccn2 |
| IUPAC Name | (4-chlorophenyl)-pyridin-2-ylmethanol |
| Molecular Weight (g/mol) | 219.67 |
| Formula Weight | 219.0451 |
| Chemical Name or Material | 4-Chlorophenyl-2-pyridinylmethanol |