CAS RN 2764892-65-9
CAS RN 2764892-65-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
2
N-Methyl Lacosamide, TRC
CAS: 2764892-65-9 Molecular Formula: C14H20N2O3 Molecular Weight (g/mol): 264.32 Synonym: (R)-2-(Acetylamino)-3-methoxy-N-methyl-N-(phenylmethyl)-propanamide,(R)-2-Acetamido-N-benzyl-3-methoxy-N-methylpropanamide IUPAC Name: (R)-2-acetamido-N-benzyl-3-methoxy-N-methylpropanamide SMILES: O=C(N(CC1=CC=CC=C1)C)[C@H](NC(C)=O)COC