Learn More
N-Methyl Lacosamide, TRC
Product Code. 30180096
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180096 50 mg 50mg
30179990 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30180096 Supplier Toronto Research Chemicals Supplier No. TRCM31521050MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2764892-65-9
Molecular Formula C14H20N2O3
Molecular Weight (g/mol) 264.32
Synonym (R)-2-(Acetylamino)-3-methoxy-N-methyl-N-(phenylmethyl)-propanamide, (R)-2-Acetamido-N-benzyl-3-methoxy-N-methylpropanamide
IUPAC Name (R)-2-acetamido-N-benzyl-3-methoxy-N-methylpropanamide
SMILES O=C(N(CC1=CC=CC=C1)C)[C@H](NC(C)=O)COC
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C14H20N2O3/c1-11(17)15-13(10-19-3)14(18)16(2)9-12-7-5-4-6-8-12/h4-8,13H,9-10H2,1-3H3,(H,15,17)/t13-/m1/s1
Formula Weight 264.15
Percent Purity >95%
Chemical Name or Material (R)-2-Acetamido-N-benzyl-3-methoxy-N-methylpropanamide
Grade HPLC