Learn More
N-Methyl Lacosamide, TRC
Specifications
Specifications
| CAS | 2764892-65-9 |
| Molecular Formula | C14H20N2O3 |
| Quantity | 2.5 mg |
| Synonym | (R)-2-(Acetylamino)-3-methoxy-N-methyl-N-(phenylmethyl)-propanamide, (R)-2-Acetamido-N-benzyl-3-methoxy-N-methylpropanamide |
| InChI Formula | InChI=1S/C14H20N2O3/c1-11(17)15-13(10-19-3)14(18)16(2)9-12-7-5-4-6-8-12/h4-8,13H,9-10H2,1-3H3,(H,15,17)/t13-/m1/s1 |
| SMILES | O=C(N(CC1=CC=CC=C1)C)[C@H](NC(C)=O)COC |
| IUPAC Name | (R)-2-acetamido-N-benzyl-3-methoxy-N-methylpropanamide |
| Molecular Weight (g/mol) | 264.32 |
| Formula Weight | 264.15 |
| Percent Purity | >95% |
| Show More |