CAS RN 27554-26-3
CAS RN 27554-26-3
IUPAC Name:
1,2-bis(6-methylheptyl) benzene-1,2-dicarboxylate
Synonyms:
Diop flexol plasticizer diop genomoll 100 hexaplas M/O corflex 880
Molecular Weight (g/mol):
390.56
Molecular Formula:
C24H38O4
InChi Key:
IJFPVINAQGWBRJ-UHFFFAOYSA-N
SMILES:
CC(C)CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC(C)C
1
–
2
of
2
results
1
Bis(6-methylheptyl) Phthalate (>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Bis(6-methylheptyl) Phthalate (>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.