Learn More
Bis(6-methylheptyl) Phthalate (>85%), TRC
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-iso-octyl ester |
| CAS | 27554-26-3 |
| Molecular Formula | C24 H38 O4 |
| Quantity | 25 mg |
| Synonym | 1,2-Benzenedicarboxylic acid, diisooctyl ester (9CI), Phthalic acid, diisooctyl ester (6CI,7CI), Corflex 880, Diisooctyl 1,2-benzenedicarboxylate, Diisooctyl o-phthalate, Diisooctyl phthalate, Hexaplas DIOP, Hexaplas M/O, Jayflex DIOP, Unem 5005 |
| InChI Formula | InChI=1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3 |
| SMILES | CCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC |
| IUPAC Name | dioctyl benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 390.56 |
| Formula Weight | 390.277 |