CAS RN 2755241-73-5
CAS RN 2755241-73-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ NR 162
Tocris Bioscience™ NR 162
Potent and selective calcium/calmodulin-dependent serine protein kinase (CASK) inhibitor