accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 27548-93-2

H3COO(R)O(S)CH3(S)OH(R)O(S)OH3CO(R)(S)OO(S)OH(S)HOCH3CH3H3C

CAS RN 27548-93-2

IUPAC Name: (1S,2S,3R,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9,15-trihydroxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
Synonyms: baccatin III
Molecular Weight (g/mol): 586.63
Molecular Formula: C31H38O11
InChi Key: OVMSOCFBDVBLFW-VHLOTGQHSA-N
SMILES: CC(=O)O[C@@H]1C2=C(C)[C@@H](O)C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)[C@@H]3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)