Learn More
Baccatine III, 95%, Thermo Scientific™
Product Code. 10367421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10367421 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10367421 Supplier Thermo Scientific Acros Supplier No. 329380050
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 27548-93-2
Melting Point 229.0°C to 234.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C31H38O11
Refractive Index 1.5455 to 1.5475
MDL Number MFCD00153921
Quantity 5 mg
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in dmso,ethanol
InChI Key OVMSOCFBDVBLFW-VHLOTGQHSA-N
SMILES CC(=O)O[C@@H]1C2=C(C)[C@@H](O)C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)[C@@H]3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C
IUPAC Name (1S,2S,3R,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9,15-trihydroxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
Molecular Weight (g/mol) 586.63
PubChem CID 71816153
Formula Weight 586.6
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Baccatine III, 95%