CAS RN 27521-34-2
CAS RN 27521-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,6alpha,17-triol (6alpha-Hydroxyethinylestradiol), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
6Alpha-Hydroxy Ethynyl Estradiol, TRC
CAS: 27521-34-2 Molecular Formula: C20H24O3 Molecular Weight (g/mol): 312.4 Synonym: 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,6alpha,17-triol,Ethinylestradiol Imp. E (EP),6alpha-Hydroxyethinylestradiol IUPAC Name: (6S,8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,6,17-triol SMILES: C[C@]12CC[C@H]3[C@@H](C[C@H](O)c4cc(O)ccc34)[C@@H]1CC[C@@]2(O)C#C