Learn More
19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,6alpha,17-triol (6alpha-Hydroxyethinylestradiol), Mikromol™
Description
19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,6alpha,17-triol (6alpha-Hydroxyethinylestradiol)
Specifications
Specifications
| Chemical Name or Material | 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,6alpha,17-triol (6alpha-Hydroxyethinylestradiol) |
| CAS | 27521-34-2 |
| Analyte or Component Names | 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,6alpha,17-triol (6alpha-Hydroxyethinylestradiol) |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C20H24O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethinylestradiol Imp. E (EP), 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,6alpha,17-triol, 6alpha-Hydroxyethinylestradiol, Ethinylestradiol Impurity E |
| SMILES | C[C@]12CC[C@H]3[C@@H](C[C@H](O)c4cc(O)ccc34)[C@@H]1CC[C@@]2(O)C#C |
| Molecular Weight (g/mol) | 312.4 |
| Quantity | 25 mg |